Would you like to log in again?
DRG® Salivary 17-OH Progesterone ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
SLV-3140
75871-388KT 213.01 USD InStock
75871-388
DRG® Salivary 17-OH Progesterone ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of active free 17‑hydroxyprogesterone (17 a-OHP) in saliva.
  • FDA cleared tests
  • Increased stability and linearity
  • Ready-to-use reagents and user-friendly assays
  • Measurement of the free (unbound) fraction of steroids
  • Excellent correlation to mass spectrometry
The steroid 17-α-Hydroxyprogesterone (17-α-OHP) is produced by both the adrenal cortex and gonads
Late onset adrenal hyperplasia:
More recently, 17-α-OHP concentrations have been utilized in the evaluation of androgenized women where late onset 21-hydroxylase is suspected. This condition is clinically very subtle and since the presentation is the same as classical polycystic ovarian disease, basal plasma 17-α-OHP concentrations, unlike classical congenital adrenal hyperplasia, are normal. The diagnosis is made by administration of an ACTH stimulation test.
The DRG Salivary 17α-OHP ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA), based on the principle of competitive binding. The microtiter wells are coated with a polyclonal antibody (rabbit) directed towards an antigenic site on the 17α-OHP molecule. Endogenous 17α-OHP of a patient sample competes with a 17α-OHP-horseradish peroxidase conjugate for binding to the coated antibody. After incubation the unbound conjugate is washed off. The amount of bound peroxidase conjugate is inversely proportional to the concentration of 17α-OHP in the sample. After addition of the substrate solution, the intensity of colour developed is inversely proportional to the concentration of 17α-OHP in the patient sample.
Even though 17-α-OHP has relatively little progestational activity, it is of intense clinical interest because it is the immediate precursor to 11-desoxycortisol (Cpd-S). Because Cpd-S is produced by 21-hydroxylation of 17-α-OHP, measurement of 17-α-OHP is a useful indirect indicator of 21-hydroxylase activity. In congenital 21-hydroxylase deficiency, the most common variety of Congenital Adrenal Hyperplasia (CAH), 17-α-OHP is secreted in abundant excess. It is moderately elevated in the 11-β-hydroxylase deficiency as well. Measurement of 17-α-OHP is therefore valuable in the initial diagnosis of CAH.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty