Would you like to log in again?
DRG® Salivary Estradiol ELISA, DRG International, Inc.
Supplier: DRG International
DRG®
SLV-4188
75871-390KT 248.33 USD InStock
75871-390
DRG® Salivary Estradiol ELISA, DRG International, Inc.
Assays ELISAs
An enzyme immunoassay for the quantitative measurement of active free estradiol, an estrogenic steroid, in saliva.
  • FDA cleared tests
  • Increased stability and linearity
  • Ready-to-use reagents and user-friendly assays
  • Measurement of the free (unbound) fraction of steroids
  • Excellent correlation to mass spectrometry
Estradiol (1,3,5(10)-estratriene-3,17β-diol; 17β-estradiol; E21) is a C18 steroid hormone with a phenolic A ring
In non-pregnant women with normal menstrual cycles, estradiol secretion follows a cyclic, biphasic pattern with the highest concentration found immediately prior to ovulation. The rising estradiol concentration is understood to exert a positive feedback influence at the level of the pituitary where it influences the secretion of the gonadotropins, follicle stimulating hormone (FSH), and luteinizing hormone (LH), which are essential for follicular maturation and ovulation, respectively. Following ovulation, estradiol levels fall rapidly until the luteal cells become active resulting in a secondary gentle rise and plateau of estradiol in the luteal phase. During pregnancy, maternal serum Estradiol levels increase considerably, to well above the pre-ovulatory peak levels and high levels are sustained throughout pregnancy.
The DRG Salivary Estradiol ELISA kit is based on the competition principle and the microplate separation. An unknown amount of Estradiol present in the sample and a fixed amount of Estradiol conjugated with horse-radish peroxidase compete for the binding sites of a polyclonal Estradiol antiserum coated onto the wells. After two hours incubation the microtiter plate is washed to stop the competition reaction. Having added the substrate solution the concentration of Estradiol is inversely proportional to the optical density measured.
This steroid hormone has a molecular weight of 272.4. It is the most potent natural Estrogen, produced mainly by the Graffian follicle of the female ovary and the placenta, and in smaller amounts by the adrenals, and the male testes. Estradiol (E2) is secreted into the blood stream where 98% of it circulates bound to sex hormone binding globulin (SHBG) and to a lesser extent to other serum proteins such as albumin. Only a small fraction circulates as free hormone or in the conjugated form. Estrogenic activity is effected via estradiol-receptor complexes which trigger the appropriate response at the nuclear level in the target sites. These sites include the follicles, uterus, breast, vagine, urethra, hypothalamus, pituitary and to a lesser extent the liver and skin.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty