Would you like to log in again?
DRG® Sperm Antibody ELISA, DRG International
Supplier: DRG International
DRG®
EIA-1826
75871-408KT 414.36 USD InStock
75871-408
DRG® Sperm Antibody ELISA, DRG International
Assays ELISAs
An enzyme immunoassay for the quantitative determination of antibodies directed against spermatozoa antigens in serum.
  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests
The Sperm Antibody ELISA (Enzyme Linked ImmunoSorbent Assay) is a solid-phase sandwich enzyme-immunoassay for the quantitative determination of anti-spermatozoa antibodies in human serum. The ELISA-plate is coated with a mix of spermatozoa proteins which are recognized by anti-spermatozoa antibodies. The samples and standards are pipetted into the wells and then incubated. During this incubation anti-spermatozoa antibodies bind to the spermatozoa proteins and are thus immobilised on the plate. After washing the enzyme conjugate, consisting of anti-human globulin antibodies covalently coupled to horseradish peroxidase, is added. After removal of the unbound conjugate by washing the horseradish peroxidase oxidizes the then added substrate TMB (3,3’,5,5’-tetramethylbenzidine) yielding a color reaction which is stopped with 0.25 M sulphuric acid (H2SO4). The extinction is measured at a wavelength of 450 nm with a microplate reader. The use of a reference measurement with a wavelength ≥550 nm is recommended.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty