Would you like to log in again?
SP Scientific miVac Sample Holder Jet Rotors for DNA and DUO Concentrators, Genevac
Supplier: Genevac
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
DRC-15CCT-012 DRC-50CCT-008 DRC-18150-006 DRC-18V40-048 DRC-28V60-014 DRC-01275-040 DRC-13100-032 DRC-12V40-078 DRC-34100-008 DRC-16100-028 DRC-20V60-035 DRC-20EPP-084 DRC-01075-068 DRC-16V35-048 DRC-05EPP-120 DRC-17115-018 DRC-15V45-048 DRC-15EPP-084
75871-530EA 1601.16 USD InStock
75871-530 75871-540 75871-532 75871-510 75871-542 75871-520 75871-534 75871-512 75871-522 75871-514 75871-536 75871-524 75871-538 75871-516 75871-526 75871-518 75871-528 75871-508
SP Scientific miVac Sample Holder Jet Rotors for DNA and DUO Concentrators, Genevac
Centrifuges Rotors
Lightweight, easily interchangeable sample holders for DNA and DUO Concentrators hold a variety of glass and plastic tubes and vials.
  • Lightweight solid aluminum construction
  • Friction fit on center spindle
  • Convenient handles
  • Hard-wearing and highly durable
  • Exceptional performance concentration
  • Wide selection available
Each miVac rotor is made to exacting specifications to assure long life, secure holding of samples and maximum heat transfer and reliability
Lightweight solid aluminum material has a very high thermal conductivity, allowing for excellent heat transfer. Precise specifications assure tubes fit tightly eliminating gaps to minimize glass breakage and further improve heat transfer. Friction fit on center spindle with convenient handles for easy installation and removal from concentrator. Rotors feature exceptional performance concentration, ranging between 40% and 200%, depending on sample format compared to rotors made from plastic.
A wide selection of sample holders are available for microcentrifuge tubes through 50ml conical tubes, as well as vials.
One of the most important factors in determining the speed of concentration is the rate at which heat (energy) is transferred to the sample. The better the heat transfer, the faster the concentration.
Ordering information: For best results, glassware should be high quality, borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty