Would you like to log in again?
SP Scientific miVac Sample Holder Jet Rotors for Quattro Concentrators, Genevac
Supplier: Genevac
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
QRC-18150-040 QRC-28V60-042 QRC-28V140-030 QRC-15EPP-200 QRC-15V60-116 QRC-01075-236 QRC-15&20EPP-200 QRC-17V60-116 QRC-35105-032 QRC-44100-014 QRC-29120-036 QRC-13100-100 QRC-15CCT-072 QRC-25150-032 QRC-01275-200 QRC-50CCT-036 QRC-20EPP-200 QRC-25V100-038 QRC-28V95-038 QRC-16100-100
75871-590EA 3220.28 USD InStock
75871-590 75871-592 75871-580 75871-550 75871-594 75871-582 75871-560 75871-552 75871-584 75871-562 75871-554 75871-576 75871-564 75871-586 75871-556 75871-578 75871-566 75871-588 75871-558 75871-548
SP Scientific miVac Sample Holder Jet Rotors for Quattro Concentrators, Genevac
Centrifuges Rotors
Lightweight, easily interchangeable sample holders for Quattro Concentrators hold a variety of glass and plastic tubes and vials.
  • Lightweight solid aluminum construction
  • Convenient handles
  • Hard-wearing and highly durable
  • Wide selection available
Each miVac rotor is made to exacting specifications to assure long life, secure holding of samples and maximum heat transfer and reliability
Lightweight solid aluminum material has a very high thermal conductivity, allowing for excellent heat transfer. Precise specifications assure tubes fit tightly eliminating gaps to minimize glass breakage and further improve heat transfer. Rotors feature convenient handles for easy installation and removal from concentrator.
A wide selection of sample holders are available for microcentrifuge tubes through 50ml conical tubes, as well as vials.
One of the most important factors in determining the speed of concentration is the rate at which heat (energy) is transferred to the sample. The better the heat transfer, the faster the concentration.
Ordering information: For best results, glassware should be high quality, borosilicate glass.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty