Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SP Scientific miVac Freeze Drying Flask Kits and Components, Genevac
Supplier: Genevac
FDA-FL150-000 FDA-FL300-000 FDA-A2932-003 FDA-IMP00-000 FDA-CONGL-003 FDA-FL600-000
75871-608EA 461.86 USD InStock
75871-608 75871-570 75871-572 75871-574 75871-610 75871-612
SP Scientific miVac Freeze Drying Flask Kits and Components, Genevac
Flasks Flat Bottom Flasks
The miVac SpeedTrap can be used in conjunction with a Super Vacuum pump.
  • Can be used with stand-alone freeze drying accessory kit or add-on flask kits
  • Freeze up to 250 ml of water or other suitable solvents
A pressure controller is recommended when samples will be freeze dried using the concentrator to assure optimal vacuum level for your process.
Samples can be placed in a rotor in a miVac concentrator or a range of flask and vial accessories are available to allow the SpeedTrap to directly accept pre-frozen samples for drying.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty