Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Dihydroxybenzoic acid

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2,5-DHBA2,5-Dihydroxybenzoic AcidGentisic acidHydroquinonecarboxylic acidDHB

D2156-25GM D2156-500GMBL D2156-100GM
75872-346EA 936 USD InStock
75872-346 75872-344 75872-342
2,5-Dihydroxybenzoic acid
2,5-Dihydroxybenzoic acid
2,5-Dihydroxybenzoic Acid, also known as Gentisic acid, is a derivative of benzoic acid and is readily oxidized and is used as an antioxidant excipient in some pharmaceutical preparations. In the laboratory, it is used as a sample matrix in matrix-assisted laser desorption/ionization (MALDI) mass spectrometry.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Formula: C₇H₆O₄
MW: 154.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty