Would you like to log in again?
BioClean™ Microflow Face Veil with Studs, Ansell
Supplier: Ansell
BioClean™
BFV06 BFV06
75873-950CS 537.64 USD InStock
75873-950 CA75873-950
BioClean™ Microflow Face Veil with Studs, Ansell
Masks Cleanroom Masks
The microflow cleanroom face veil features a fully enclosed malleable nose-band for a good fit and studs for a secure fastening to hood.
  • Ultrasonically sealed seams
  • Fully enclosed malleable nose-band
  • Studs on either side for attaching to cleanroom hoods
  • Disposable, single-use only
  • Latex-free
The face veil features ultrasonically sealed edges to reduce the risk of contamination in controlled environments. ISO Class 4 cleanroom compatible. Overall garment dimensions are 375 mm in length by 330 mm wide.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty