Would you like to log in again?
COLUMN CGNT BIDENTATE C8 4UM 100A 75X3MM
Supplier: MicroSolv Technology Corp
HPLC Column Cogent Bidentate C8 (Octyl) 75mm x 3.0mm ID^ HPLC Column, Phase type: Cogent Bidentate C8 (Octyl), Dimensions: 75mm x 3.0mm ID, Particle size: 4um, Pore size: 100A, Surface Area: 390 m2/g, Carbon Load: 13-14%, pH Range: 2.0-8.0, Less Retentive
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CGNT BIDENTATE C8 4UM 100A 75X3MM 75874-062 40008-75P-3 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty