Would you like to log in again?
COLUMN COGENT UDA(WCX) 4UM 100A 75X30MM
Supplier: MicroSolv Technology Corp
Cogent UDA (wcx) HPLC column 75mm x 30mm ID^ Cogent UDA (wcx), HPLC column, Undecanoic Acid, Dimensions: 75mm x 30mm ID, Particle size: 4um, Pore size: 100A, Surface Area: 390 m2/g, Carbon Load: 14-15%, pH Range: 2.0-8.0, Maximum temperature: 80 deg CWhic
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN COGENT UDA(WCX) 4UM 100A 75X30MM 75874-198 40031-P30-75 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty