Would you like to log in again?
COLUMN CGNT PNYL HYDE 4UM 100A 150X30MM
Supplier: MicroSolv Technology Corp
HPLC column Cogent Phenyl Hydride^ HPLC column, Phase type: Cogent Phenyl Hydride, Dimensions: 150mm x 30mm ID, Preparative Column, Particle size: 4um, Pore size: 100A, Surface Area: 390 m2/g, Carbon Load: 10-12%, pH Range: 2.0-8.0, Maximum temperature: 8
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
COLUMN CGNT PNYL HYDE 4UM 100A 150X30MM 75874-226 69020-P30-150 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty