Would you like to log in again?
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
Supplier: Eppendorf
Eppendorf Tubes®
0030119401 0030119479 0030119380 0030119452 0030119479 0030119401 0030119487 0030119487 0030119380 0030119460 0030119460 0030119606
89429-308EA 89.79 USD InStock
CA89429-308 CA89429-306 89429-308 CA10017-732 89429-310 CA89429-312 75874-634 89429-312 CA89429-310 89429-306 89429-318 76019-020
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
Tubes Microcentrifuge Tubes
Eppendorf Tubes® with snap caps are an ideal choice when working with medium-sized sample volumes (0.5 to 5.0 ml). Designed for user-friendly handling, broad temperature ranges, centrifugation safety (up to 25,000×g), and sample recovery.
  • Designed for ease of use and user-friendly handling during sample preparation and processing
  • Suitable for a wide range of temperatures, ensuring adaptability to various experimental conditions
  • Specifically designed to enhance sample recovery, preserving the integrity of samples
  • Available in Eppendorf Quality, PCR Clean, Biopur®, Sterile, and Forensic DNA Grade quality for tested purity
Explore the versatility of Eppendorf 5.0 ml tubes, available in both colorless and amber options. Tailor the selection with various certified purities, ensuring precise and reliable sample recoveries. Experience the flexibility these tubes provide, allowing selection of the ideal combination for specific research needs.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Certifications: Certificate of Quality/Conformity, Lot-specific certificates for the purity grades PCR clean, Biopur, Sterile®, Forensic DNA Grade can be downloaded from the Eppendorf website.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty