Would you like to log in again?
Eppendorf Tubes® 5.0 ml
Supplier: Eppendorf
0030108302 0030119401 0030108302 0030119479 0030122313 0030119380 0030122321 0030119452 0030108310 0030119479 0030119401 022365001 0030119487 0030119487 0030122330 0030119380 0030108310 0030119460 0030119460 0030122305
89429-308EA 89.79 USD InStock
CA89429-308 CA89429-306 89429-308 75874-640 89429-314 CA89429-312 89429-312 CA89429-332 CA89429-310 89429-306 76313-962 CA89429-316 89429-318 CA89429-314 89429-316 CA10017-732 89429-310 75874-636 75874-634 75874-638
Eppendorf Tubes® 5.0 ml
Tubes Test Tubes
Eppendorf tubes 5.0 ml represents the perfect option for working with medium sized sample volumes. It fills the gap between existing tube formats and enables the simple and safe processing of sample volumes up to 5.0 ml.
  • Ergonomic operation
  • Snap cap and screw cap
  • Certified purity
  • Less storage space needed
  • Easier sample access lowers the risk of contamination
Applications include preparation of mastermixes and buffers, convenient and safe cell culture applications, safe cell and tissue lysis, and higher yields in DNA isolation, especially with samples with limited quantity of source material.
The tubes provide maximum safety and stability for centrifugation up to 25000 ×g with fast and efficient protocols. These conical shape is compatible with accessories for conical 15 ml tubes, so many existing adapters and racks can be used. The 5.0 ml snap cap tubes are available in Eppendorf LoBind® material for maximum recovery of valuable samples.
Snap caps allow simple, practical and ergonomic singlehand operation, while screw caps have flattened and grooved sides which provide optimal grip and user-friendly handling. The tubes also have a broad operating temperature range: Screw cap tubes: −86 to 100 °C. Snap cap tubes: −86 to 80°C (when using the clip ≤100 °C). They are made from an exceptionally high-quality, transparent polypropylene, free of plasticizers, biocides or mold release agents, for reliable test results. Available in lot specific certified Eppendorf Quality, PCR clean, Sterile and Eppendorf Biopur® purity grades.
Ordering information: The 5 ml tube packages include 400 tubes (2 packages with 2 bags of 100 each), 2 white racks (with 12 places each), and 8 universal adapters for rotors with bore holes for 15 ml conical tubes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty