Would you like to log in again?
Sealing Mats for Deep Well Plates, Eppendorf
Supplier: Eppendorf
0030127960 0030127978
75874-653PK 316.59 USD InStock
75874-653 75874-655
Sealing Mats for Deep Well Plates, Eppendorf
Seals Microplate Seals
Eppendorf sealing mats for deep well plates.
  • Autoclavable
  • Sample storage and preparation
  • Aluminum
Can be pierced. Use to protect light-sensitive samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty