Would you like to log in again?
PCR Tube Strips and Cap Strips, Eppendorf
Supplier: Eppendorf
0030124820 0030124839 0030124847 0030124812 0030124804
75874-708CS 103.31 USD InStock
75874-708 75874-702 75874-710 75874-706 75874-704
PCR Tube Strips and Cap Strips, Eppendorf
Tubes PCR Tubes
Original Eppendorf PCR tubes are manufactured according to the highest quality standards.
  • Eight reaction tubes in strip-format
  • Ideal for small sample volumes
  • Easily adaptable for automation
  • Sealable using flat or domed strip-lids
  • PCR with medium sample throughput
  • Flat Cap Strips are suitable for real-time PCR
Eppendorf cap strips provide easy and rapid sealing of Eppendorf twin.tec® PCR plates, PCR strips and other standard multi-well plates. Strips with eight microcaps for 0.1 and 0.2 ml wells. Caps are autoclavable.
These thin-walled polypropylene tubes ensure efficient and homogenous heat transfer. The purity grade PCR Clean is safeguarded by closed bags being inside an outer carton.
Certified free from human DNA, DNase, RNase and PCR inhibitors.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty