Would you like to log in again?
Eppendorf® PCR Tube and Plate Storage Rack
Supplier: Eppendorf
0030124545 0030124545 0030124545
89135-368CS 327.66 USD InStock
CA89135-368 89135-368 75874-714
Eppendorf® PCR Tube and Plate Storage Rack
Racks Tube Racks
PCR Racks are designed for handling and storing tubes and plates
Racks accommodate 0.1, 0.2 and 0.5mL PCR tubes, PCR tube strips, and 96-Well PCR plates. They are stackable and can be stored at temperatures as low as –90°C (–130°F). Autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty