Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

New Brunswick™ Innova® 43/43R Console Incubator Shakers

Supplier: Eppendorf
New Brunswick™ Innova®
M1320-0010 M1320-0014 M1320-0010 M1320-0004 M1320-0004 M1320-0000 M1320-0000 M1320-0014
75874-892EA 25998.51 USD InStock
75874-892 CA75874-892 CA75874-520 75874-520 CA75874-538 CA75874-516 75874-516 75874-538
New Brunswick™ Innova® 43/43R Console Incubator Shakers
Shakers and Mixers Shaking Incubators
The Eppendorf Innova 43 and 43R (refrigerated) floor-standing incubator shakers are the ideal solution for the precise handling of large culture volumes. The triple-eccentric counterbalanced drive is designed to support heavy workloads - the design has stood the test of time for over 70 years and continues to enable scientists today.

  • Applicational flexibility: Accommodates flasks up to 6 L, sticky pad platform for flat bottom vessels, orbits 25 or 51 mm (1 or 2"), optional: photosynthetic light bank and humidity monitoring
  • Saves money: No internal HEPA-filter that must be replaced annually for optimal performance
  • Reliable 24/7 with high loads: Proven and tested Innova triple eccentric counterbalanced drive (three weight-supporting shafts)
  • Easy cleaning: Spill diverter and drainable built-in spill pan with rounded corners protect the drive from accidental spills (can be used as a water reservoir to reduce sample evaporation); airflow UV-germicidal light optional
  • Hands-free door operation: Foot pedal release with easy-to-open and close lid
  • Ergonomical and safe interface handling: Top mounted controls and display
  • Programmability: Continuous, timed, or multi-step programming (15 steps), store up to four programs
  • Serviceability: Built-in castors to move shaker easily, qualified lab shaker service available

The Innova 43 and 43R (refrigerated) shakers are the perfect choice for handling large volumes of cultures with precision and ease. The top loading design allows for vertical placement of the flasks. A pedal operated lid and a window in the sidewall overlooking the samples are examples of why our customers continue to trust our designs. The cast iron triple-eccentric counterbalanced drive is specifically designed to support high-speed applications and heavy workloads - the design has stood the test of time for over 70 years and continues to enable scientists today. The simple user interface allows for easy programming of speed, temperature, time and the VisioNize box can be added to enhance monitoring and adds notifications functionality.

4 Stored programs.

Ordering information: Options and accessories include: UV germicidal light, humidity monitoring, documentation and monitoring with notifications with VisioNize® Lab Suite and more.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty