Would you like to log in again?
Heat Sealer, S200
Supplier: Eppendorf
5392000013 5392000013
75874-928EA 6322.33 USD InStock
75874-928 CA75874-928
Heat Sealer, S200
Heat Sealers Microplate Heat Sealers
The Eppendorf HeatSealer S200 offers flexibility regarding the sealing temperature, sealing time and plate formats.
  • Hermetic sealing of multiwell plates
  • Fixed sealing temperature for reproducible results
  • Eliminates evaporation in PCR, reducing cross contamination
  • Seals 96- and 384-well plates
  • Compact design, portable device
  • Integrated thermostat prevents overheating
  • Improved mechanics for even easier sealing
  • PCR Plate Adapter included
The large handle allows for high sealing pressures.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty