Would you like to log in again?
Refrigerated Microcentrifuges, 5430 R
Supplier: Eppendorf
022620623 022620667 022620656 022620601 022620690 022620634 022620645 022620638 022620601 022620623 022620642 022620634 022620666 022620689 022620690 022620642 022620638 022620678 022620645 022620625 022620663 022620603 022620667 022620659 022620678 022620656 022620665
97027-862EA 16755.67 USD InStock
CA10030-958 97027-862 10030-960 97027-860 97027-858 75875-088 CA97027-866 97027-856 97027-854 CA97027-862 89422-676 CA97027-848 10030-958 97027-852 CA10030-960 CA97027-860 CA97027-858 97027-848 CA97027-856 CA97027-854 97027-866 75875-650 CA97027-852 97027-864 75875-090 75875-092 75875-094
Refrigerated Microcentrifuges, 5430 R
Centrifuges Microcentrifuges
The cross-over refrigerated Eppendorf Centrifuge 5430 R combines the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, it can accommodate rotors for microplates and 15/50 ml conical tubes as well as Eppendorf tubes and PCR strips.
  • Remarkable versatility with 12 different rotors
  • Soft-touch one-finger lid closure for ergonomic operation
  • Menu-driven, multi lingual operation menu (English, German, French, Spanish) with large backlit display
  • 5 program keys for easy access to routine programs
  • Saves up to 50 user-defined programs
  • Automatic rotor recognition and imbalance detection for maximized operational safety
  • Eppendorf QuickLock® - System for quickly opening and closing the rotor lid
The compressor technology reduces vibrations and protects your samples. Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool. FastTemp pro® function for programmable pre-cooling according to day and time. ECO shut-off engages after eight hours of nonuse to reduce energy consumption (37% overnight) and extend compressor life (can be deactivated in menu) and built-in condensation drain to eliminate water accumulation.
Centrifuge 5430 R combines the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument. The high-speed centrifuge offers centrifuge rotors for Eppendorf tubes and PCR strips, but also rotors for microplates and 15 or 50 ml conical tubes.
Additional Eppendorf QuickLock® centrifuge rotors are now available in the rotor program, including 48-place rotors (1.5 ml or 2.0 ml), a 16-place rotor for Eppendorf Tubes 5.0 ml and a swing-bucket rotor (24×1.5 ml or 2.0 ml). With its unmatched versatility, Centrifuge 5430 R is the flagship product among the Eppendorf microcentrifuge portfolio.
Available as keypad or knob variant.
Certifications: UL Listed, Certificate of Quality/Conformity (ISO 9001, ISO 13485, ISO 14001).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty