Would you like to log in again?
Benchtop Centrifuge, 5702/5702 R/5702 RH
Supplier: Eppendorf
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
022629916 022628102 022626213 022628113 022626205 022628113 022628001 022628102 022629921 022628124 022629888 022628023 022626213 022628124 022626205 022629905 022626256 022628012 022626001 022629883 022626001 022626108 022628012 022628001 022626230 022628023
89207-270EA 12468.51 USD InStock
CA47745-374 89207-270 CA80076-418 CA89207-262 CA89207-264 CA89207-266 47743-472 89203-054 89207-272 CA89207-270 80076-418 80081-164 CA89207-272 47745-374 80081-166 89207-268 75875-104 CA47743-472 89207-266 89207-264 89207-262 97007-364 97007-366 97007-362 80076-420 CA89207-268
Benchtop Centrifuge, 5702/5702 R/5702 RH
Centrifuges Benchtop Centrifuges
The range of Eppendorf 5702 general purpose, low-speed centrifuges has been designed specifically to meet the needs of mid-throughput clinical research and cell culture laboratories. These centrifuges are very compact, so that they fit easily on the laboratory bench, as well as extremely quiet.
  • Very compact footprint to fit on every laboratory bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • 'At set RPM' function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety
In addition to the above features centrifuges 5702 R and 5702 RH feature the following:
Fast temprature function for fast precooling.
Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool.
ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life.
Two program keys to store routine programs.
Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH).
For in-vitro diagnostic use. The Centrifuge 5702/5702 R/5702 RH is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose.
Make your laboratory environment a better place to work with the 5702 family of low-speed centrifuges that are quiet, user-friendly and so compact that you can fit one comfortably on every lab bench. The low access height allows simple loading and unloading of samples.
The 5702 family offers three different models to suit your laboratory or your application. Centrifuge 5702, which is suitable for standard applications, the refrigerated Centrifuge 5702 R for heat sensitive samples, and the actively heated and refrigerated Centrifuge 5702 RH, which is well suited for working with living specimens, for example in cell culture.
Certifications: UL listed, certificate of quality/conformity (ISO 9001, ISO 13485, ISO 14001).
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty