Would you like to log in again?
Rotors for Centrifuges, 5920 R
Supplier: Eppendorf
5895101003 5895118003 5895190006 5895100007 5895117007
75875-252EA 5467.12 USD InStock
75875-252 75875-254 75874-312 75875-256 75874-314
Rotors for Centrifuges, 5920 R
Centrifuges Rotors
Eppendorf Rotors for centrifuges 5920 R - precision in refrigerated large-capacity centrifugation. Tailor your experiments with varied capacities and speeds. Accessorize with versatile buckets and adaptors for efficient sample processing. Ideal for high-throughput applications, ensuring accuracy and reproducibility.
  • Precision-engineered for optimal sample separation
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 min)
  • Accessories (adaptors and buckets) further customize your centrifuge setup
  • Aerosol-tight options available
Experience precision in refrigerated large-capacity centrifugation with Eppendorf rotors designed for centrifuges 5920 R. These rotors offer a range of capacities and speeds, providing flexibility for various sample processing needs. Accessorize with versatile buckets and adaptors, enhancing the adaptability of your centrifuge setup. Ideal for high-throughput applications, the centrifuges 5920 R is further optimized with these precision-engineered rotors, ensuring accuracy and reproducibility in every spin. Explore the range to elevate your refrigerated large-capacity centrifugation experience.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty