Would you like to log in again?
Quartz Ground and Polished Plates, Technical Glass Products (TGP)
Supplier: Technical Glass Products (TGP)
RQP6-6-16 RQP1.5-1.5-16 RQP1.5-1.5-14 RQP1.5-1.5-18 RQP5-5-18 RQP1-1-16 RQP3-3-14 RQP1-1-18 RQP5-5-16 RQP2-2-18 RQP2--2-16 RQP4-4-16 RQP3-3-16 RQP1-1-14 RQP3-3-18 RQP4-4-18 RQP2-2-14 RQP6-6-18 RQP5-5-14 RQP4-4-14 RQP6-6-14
75875-424EA 133.78 USD InStock
75875-424 75875-402 75875-412 75875-434 75875-404 75875-426 75875-436 75875-414 75875-406 75875-428 75875-416 75875-438 75875-408 75875-418 75875-440 75875-420 75875-430 75875-400 75875-422 75875-410 75875-432
Quartz Ground and Polished Plates, Technical Glass Products (TGP)
Optical Windows
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These plates provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Ground and polished plates manufactured from this material are invaluable in research and industry applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty