Would you like to log in again?
Quartz Ground and Polished Discs, Technical Glass Products (TGP)
Supplier: Technical Glass Products (TGP)
RQD3-116 RQD4-116 RQD1.5-18 RQD3-14 RQD4-18 RQD1.5-116 RQD1-16 RQD2-18 RQD4-14 RQD2-14 RQD1-18 RQD1-14 RQD3-18 RQD2-116
75875-468EA 149.64 USD InStock
75875-468 75875-446 75875-456 75875-448 75875-458 75875-460 75875-462 75875-450 75875-464 75875-442 75875-452 75875-466 75875-444 75875-454
Quartz Ground and Polished Discs, Technical Glass Products (TGP)
Optical Windows
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These discs provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Ground and polished discs manufactured from this material are invaluable in research and industry applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty