Would you like to log in again?
Quartz Microscope Slides, Technical Glass Products
Supplier: Technical Glass Products (TGP)
RQMS1-3-1 RQMS1-2-1 RQMS1-1-1
75875-470EA 24.62 USD InStock
75875-470 75875-472 75875-474
Quartz Microscope Slides, Technical Glass Products
Slides Microscope Slides
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These microscope slides provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Ground and polished slides manufactured from this material are invaluable in research and industry applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty