Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quartz Cover Slips, Technical Glass Products

Supplier: Technical Glass Products (TGP)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

RQCS1-1 RQCS1
75875-476EA 24.22 USD InStock
75875-476 75875-478
Quartz Cover Slips, Technical Glass Products
Cover Glasses
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%

Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These cover slips provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.

Cover slips manufactured from this material are invaluable in research and industry applications.

Grade: GE124

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty