Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

New Brunswick Innova® 40/40R Benchtop Orbital Shakers

Supplier: Eppendorf
New Brunswick Innova®
M1299-0094 M1299-0094 M1299-0080 M1299-0090 M1299-0084 M1299-0090 M1299-0084 M1299-0080
75874-502EA 13411.14 USD InStock
CA75874-408 75874-502 75874-534 CA75875-636 75874-408 75875-636 CA75874-534 CA75874-502
New Brunswick Innova® 40/40R Benchtop Orbital Shakers
Shakers and Mixers Orbital Shakers
The Innova 40/40R is an easy-to-program orbital shaker that comes with the famous Innova triple eccentric drive to provide 24/7 reliable operation for decades.

  • Quiet operation provides a favorable work enviornment
  • Built to perform and last: Innova triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble-free operation for years
  • 1.9 and 2.54 cm (3/4 and 1") orbits are available to meet application needs
  • Save space on the laboratory bench: Small footprint of 55.9×76.2 cm/22×30" (W×D) that even works on narrow benches
  • Wide temperature range Innova 40: 10 °C above ambient to 80 °C. (Innova 40R: 20 °C below ambient to 80 °C)
  • Minimizes splashing and mechanical damage: Acceleration/deceleration circuit prevents sudden starts and stops
  • Easy to move: 60/79 kg weight (incubated/refrigerated version)
  • Versatile 46×46 cm (18×18") accessory platforms (sold separately) accommodates flasks up to 3 L as well as test tube racks and microplate holders

The Innova 40 and 40R (refrigerated) benchtop shakers provide years of dependable operation. This is the perfect shaker to begin working on small-scale projects with a view to increase volume or throughput. The self-supporting transparent lid allows samples to be in view all the time. The ability to shake flasks up to 3 L in such a compact benchtop unit provides clear scope to upscale and options include a drip pan and remote alarm. The simple user interface allows for easy programming of speed, temperature and time. The VisioNize box can be added to enhance monitoring and adds notifications functionality. Regular servicing can be carried out in-situ, ensuring minimized downtime.

Drive mechanism: Triple-eccentric counterbalanced drive
Stackable: Non stackable
Memory: Non-volatile with automatic power failure restart
Steps per program: 15
Motor type: Solid state, DC brushless motor
Stored programs: 4

Certifications: Meets cETL, UL, CSA and CE standards.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty