Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
LaboPlast®/SteriPlast® DispoLadle, Bürkle
Supplier: Bürkle
5393-6641 5393-6650 5393-6640 5393-6651
75876-060PK 222.53 USD InStock
75876-060 75876-066 75876-064 75876-062
LaboPlast®/SteriPlast® DispoLadle, Bürkle
Scoops
DispoLadle is a sampling scoop specially designed for disposable use
  • 200 mL (6.76 oz.) scoop
  • External diameter of 92 mm (3.62")
  • Ergonomic handle
  • HDPE construction
  • Cleanroom manufactured, individually packaged
  • Optionally sterilized by gamma rays
The scaled scoop with embossing can be used for measuring, and displays contents in increments of 10mL (0.34oz.) up to 200mL (6.76oz.). An ergonomic handle and a spout on both sides for right- or left-handed operation make handling simple and easy.
Packaging: 20 DispoLadles per package.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty