Would you like to log in again?
SteriPlast® Scraper for Foodstuffs, Bürkle
Supplier: Bürkle
SteriPlast®
5378-3030
75876-068PK 43.66 USD InStock
75876-068
SteriPlast® Scraper for Foodstuffs, Bürkle
Scrapers
The SteriPlast® Scraper for Foodstuffs, made of blue polystyrene, is produced in a class 7 cleanroom (10,000), individually packaged for disposable use, and then sterilized by gamma rays.
  • Can be used as part of HACCP/IFS foreign object management
  • Ideally suited for the food, animal feed, and pharmaceutical industries
Sturdy and convenient scraper for disposable use. The scraper is suitable for scraping off, smoothing, or cleaning, as well as removing residue.
Packaging: 10 scrapers per package.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty