Would you like to log in again?
LaboPlast®/SteriPlast® Spoon Spatula, Bürkle
Supplier: Bürkle
LaboPlast®
5378-0031 5378-1031
75876-076PK 103.66 USD InStock
75876-076 75876-074
LaboPlast®/SteriPlast® Spoon Spatula, Bürkle
Spatulas
The spoon spatula is a double spatula with a 0.5 ml (0.02 oz.) spoon and a 17 mm (0.67") spatula blade.
  • Polystyrene construction
  • Production, assembly, and packaging according to Cleanroom Class 7
  • Complies with EU food requirements and FDA regulations
  • Optionally packaged individually and sterilized by gamma rays
The spoon spatula has many different uses in lab applications. It may be used as a sampling spoon, for measuring or transferring small amounts, as a scraper, or for mixing and stirring. Specially designed for disposable use, it is resistant to chemicals and corrosion-proof.
Packaging: Cleanroom manufactured, Spoon Spatulas are available in two packaging options. LaboPlast® Spoon Spatulas are supplied in a large package with 100 pieces. SteriPlast® Spoon Spatulas feature 100 pieces packaged individually and sterilized by gamma rays.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty