Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LaboPlast® Rotating Pipette Stand, Bürkle

Supplier: Bürkle
LaboPlast®
9622-0001 7118-2001 9630-0090 9630-0100
75876-082EA 27.86 USD InStock
75876-082 75846-220 75876-088 75876-086
LaboPlast® Rotating Pipette Stand, Bürkle
Pipette Stands
The LaboPlast® Rotating Pipette Stand is designed as a frame for pipettes with two rotating perforated plates and a drip pan

  • Polypropylene, chrome-plated steel construction
  • Adjustable height up to 450mm (17.72")
  • Accommodates up to 40 pipettes, with a maximum diameter of 18mm (0.71")

The stand will accommodate all measuring and full pipettes up to a maximum diameter of 18mm (0.71"). The drip pan protects the tips of the pipettes and collects liquid as it drips off. A very sturdy pedestal ensures absolute safety against overturning. No clamping screws are necessary due to an innovative magnet concept. The height of the holding plates can be adjusted easily with one hand.

The holders are suitable for up to 40 pipettes of different sizes.

Packaging: One pipette stand per package.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty