Would you like to log in again?
Rejuvenator® Polyurethane Squared Surface Runners, Wearwell®
Supplier: WEARWELL
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Rejuvenator®
502.58X3X5CBK 502.58X2X5EFBK 502.58X2X5CBK 502.58X2X5EMBK 502.58X3X5EMBK 502.58X3X5EFBK
75876-306EA 322.06 USD InStock
75876-306 75876-338 75876-238 75876-336 75876-340 75876-240
Rejuvenator® Polyurethane Squared Surface Runners, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Rejuvenator® runners deliver unmatched anti-fatigue benefits and long-term performance.
  • 100% urethane with thick outer wear layer
  • Deep positive lock keeps seams secure
  • Quickly form endless lengths
  • Molded-in safety bevels
  • Meets USP 797 compliance standards
  • Ideally suited for dry environments
Rejuvenator® Runners provide the option to create 2' or 3' wide runs of ergonomic matting. The mats are available in Ends and Centers, allowing users to create the specific length of run. The molded-in ramps reduce installation time and costs because there is no edging to attach.
Certifications: ASTM F1677. Dry: COF = 0.75. Wet: COF = 0.28.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty