Would you like to log in again?
Single Mouse Metabolic Cage, Tecniplast®
Supplier: Tecniplast
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3600M021
75876-736EA 1341.45 USD InStock
75876-736
Single Mouse Metabolic Cage, Tecniplast®
Animal Supplies Animal Cages
This metabolic cage is used for the efficient collection of urine and feces from a single mouse
The smaller chamber volume and collection tube design minimize evaporation. The funnel and cone design, with a steeper angle, separates urine and feces into separate collection tubes. Non-wetting funnel and cone are made of polymethylpentene (PMP) plastic to ensure immediate and complete separation of urine and feces. Cage is self-standing on a benchtop or shelf rack. Support plate 73521-334 allows the cage to fit the 12-cage support racks.
Ordering information: Includes complete assembly and use instructions, replacement parts list, and warranty card.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty