Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Desmin IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3255B 3255A
75876-888BX 658.84 USD InStock
75876-888 75876-886
Desmin IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Desmin control slides contain sections of positive staining leiomyoma uterus and negative staining tonsil.

  • Tissue type: (+) Leiomyoma uterus / (-) Tonsil
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply desmin control slides are for the positive immunohistochemical staining of Desmin, a protein found in normal smooth, skeletal, and cardiac muscle cells and expressed in leiomyoma, leiomyosarcoma, rhabdomyoma, and rhabdomyosarcoma tumors.

Staining Results:
Desmin positive expression will stain brown cytoplasmic staining.
Tonsil will stain negative.
Nuclei will stain blue.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty