Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gram, Multi-Tissue, Artificial Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4256A 4256B
75877-026BX 511.66 USD InStock
75877-026 75877-024
Gram, Multi-Tissue, Artificial Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Gram, multi-tissue, artificial control slides contain sections of gram positive staining rat lung, gram negative staining rat lung and a section of human lung nonreactive to organisms.

  • Tissue Type: (+) Gram Pos Rat Lung (+) Gram Neg Rat Lung / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply Gram, Multi-tissue, artificial control slides are for the positive histochemical staining of gram positive and gram negative bacteria in separate tissue sections.

Staining Results:
Gram negative bacteria will stain red.
Gram positive bacteria will stain blue/violet.
Nuclei will stain red.
Background tissue will stain yellow.
Nonreactive lung will stain negative for gram positive/negative bacteria.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty