Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fungus, GMS, Candida sp., Artificial Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4230B 4230A
75877-154BX 511.66 USD InStock
75877-154 75877-152
Fungus, GMS, Candida sp., Artificial Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Fungus, GMS, Candida sp., artificial control slides contain sections of positive staining rat lung and negative staining human lung.

  • Tissue Type: (+) Rat Lung / (-) Lung
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply Fungus, GMS, Candida sp., artificial control slides are for the positive histochemical staining of Candida sp

Staining Results:
Candida sp. will stain sharply outlined in black.
Background will stain green.
Negative lung will stain negative for fungus.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

in tissue sections.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty