Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Helicobacter sp. Histo Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
4276B 4276A
75877-304BX 97.98 USD InStock
75877-304 75877-306
Helicobacter sp. Histo Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Helicobacter sp. control slides contain sections of positive staining gastrointestinal tract and negative staining lung.

  • Tissue Type: (+) GI Tract
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • Histo slides are cut at 4 µm, unless otherwise specified
  • Standard tissue placement is mid-high on slide

Newcomer supply Helicobacter sp. Control slides are for the positive histochemical staining of Helicobacter sp., a spiral shaped bacterium associated with stomach inflammation and implicated in the development of gastric malignancy, peptic ulcers, and chronic gastritis.

Staining Results:
Helicobacter sp. will stain dark brown to black.
Background will stain golden brown.
Lung will stain negative for Helicobacter sp.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of Histo control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: In human tissue unless otherwise indicated. Histopathology control slides are not quality controlled for IHC methods, as such, we cannot guarantee antigenicity.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty