Would you like to log in again?
BioClean™ P-ZERO™ Sterile Polychloroprene Gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioClean P-Zero™ BioClean™
BPZS60 BPZS65 BPZS85 BPZS70 BPZS75 BPZS80 BPZS65 BPZS75 BPZS90 BPZS60 BPZS70 BPZS80 BPZS85 BPZS90
75877-390CS 639.34 USD InStock
CA75877-386 CA75877-398 75877-390 CA75877-384 75877-396 CA75877-396 75877-384 75877-398 75877-386 CA75877-394 75877-392 CA75877-392 75877-394 CA75877-390
BioClean™ P-ZERO™ Sterile Polychloroprene Gloves, Ansell
Gloves
BioClean™ P-Zero™ Sterile Polychloroprene Gloves offer excellent sensitivity and cytotoxic drug protection.
  • Chemical resistant
  • Excellent tactility
  • Double-donnable
  • Textured for improved grip
  • Non-particulating EasyTear™ packaging
Providing exceptional grip, sensitivity, and cytotoxic protection, BioClean™ P-Zero™ Sterile Polychloroprene Gloves have excellent antistatic properties and offer both chemical protection and outstanding tactility.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Directive 89/686/EEC.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of Glove Standard EN420:2003 and EN374-1, 2 & 3:2003.
Physical properties comply with European Medical Glove Standard EN455-2:2009.
Tested against permeation standard ASTM D6978-05 for cytotoxic drugs.
Sterilized in accordance with ISO 11137-1:2006.ISO Class 4 compatible.
Packaging: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 20 outer bags per carton liner; one carton liner per carton (200 pairs).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty