Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: MP Biomedicals
0855550
75877-470EA 511.6 USD InStock
75877-470
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated goat IgG fraction to mouse IgG (whole molecule) and buffer salts. Anti-Mouse IgG is developed in goat using purified mouse IgG as the immunogen. Affinity isolation removes essentially all goat serum proteins, including immunoglobulins which do not specifically bind to mouse IgG.

This product is suitable for use as a reagent in enzyme immunoassays (EIA), cell and tissue staining (for light microscopy), cell and tissue labeling (for electron microscopy), and blot immunostaining. (Note: F(ab')2 fragments are recommended for staining of cells or tissues which contain Fc receptors. Affinity purified antibodies or their fragments are recommended to avoid non-specific binding from inherent antibodies of host animals).

Antibody and highly purified HRP (Rz>3.0) are conjugated under defined conditions to obtain optimally labeled product. Conjugated protein is purified by salt fractionation. The product is dialyzed into 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3, adjusted to standard titer, filtered through a 0.22 µm filter, vialed and lyophilized. No preservative has been added.

The total protein is measured using the Biuret procedure with bovine albumin as the standard. To ensure that the product titer falls within the required range, antibody titer is standardized by microtiter plate ELISA with mouse IgG. The product is tested for purity and specificity at final concentration by immunoelectrophoresis. The antibody is predominantly goat IgG; no trace of albumin is detected.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty