Would you like to log in again?
Extracta DBS, Quantabio
Supplier: Quantabio
Extracta™
95171-500 95171-010 95171-010 95171-500
75877-506EA 762 USD InStock
CA75877-508 CA75877-506 75877-506 75877-508
Extracta DBS, Quantabio
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Extracta DBS is a rapid and easy to use extractions reagent optimized for the extraction of DNA from dried blood spot punches
  • Optimized for DNA extraction from dried blood spot punches
  • Single reagent for PCR-ready DNA in less than 30 minutes
  • Maximized assay sensitivity, lower Ct’s, when combined with Quantabio ToughMix
  • Compatible with high-throughput automation for PCR, qPCR and NGS applications
  • Reliable, Consistent Lot-to-Lot Performance
Extracta DBS is a ready-to-use DNA extraction reagent for rapid and efficient recovery of PCR-ready DNA from dried blood spots (DBS) on Guthrie cards or Whatman 903 filter paper. This patented single-solution process produces DNA eluates that are substantially free of PCR inhibitors and compatible with a variety of end-point PCR, real-time PCR and Next Generation Sequencing (NGS) or Sanger Sequencing (1-3) reagents. Application of Extracta DBS with PerfeCta® qPCR ToughMix® or PerfeCta® MultiPlex qPCR ToughMix enables accurate and reproducible quantification of DNA sequences in blood using TaqMan® hydrolysis probe real-time qPCR.
The resulting extract is immediately usable for downstream applications such as real-time PCR.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty