Would you like to log in again?
BioClean™ Sterile Neoprene Gloves, Ansell
Supplier: Ansell
BioClean Suprene™
BSNS-85 BSNS-85 BSNS-60 BSNS-75 BSNS-70 BSNS-65 BSNS-60 BSNS-75 BSNS-65 BSNS-80 BSNS-80 BSNS-70 BSNS-90 BSNS-90
75877-712CS 1233.61 USD InStock
CA75877-546 75877-712 CA75877-712 75877-546 CA75877-710 75877-548 75877-710 CA75877-552 CA75877-708 75877-708 CA75877-548 75877-550 CA75877-550 75877-552
BioClean™ Sterile Neoprene Gloves, Ansell
Gloves
BioClean Suprene™ sterile neoprene cleanroom gloves are resistant to a wide range of chemicals. Their excellent strength and stretch these gloves are the ideal choice for use within isolators.
  • Double-donnable with good grip
  • Strong and durable
  • Textured fingers and palms
  • Non-particulating EasyTear™ packaging
  • Powder-free, Latex-free to eliminate Type 1 latex allergies
The hand specific shape and soft neoprene ensure a comfortable and ergonomic fit. Suitable for use in Class 10, ISO 4 or M2.5 and EU GMP Grade A cleanrooms.
Beaded cuff and textured surface for increased tactility and also it comes with excellent ESD properties.
Certifications: In accordance with ASTM D6978. Complies with EN 374-1, 2 & 3:2003 and EN 455-2:2009 glove standards. ISO 9001: 2008 facility. Sterilized in accordance with ISO 11137-1:2006. Conforms to Category 3.Processed in an ISO Class 4 environment. CE 0120 marked.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty