Would you like to log in again?
PTFE Bench-Scale Clean-In-Place Sprayer, Ace Glass
Supplier: Ace Glass
6446-01 6446-10
75877-554EA 857.07 USD InStock
75877-554 75877-556
PTFE Bench-Scale Clean-In-Place Sprayer, Ace Glass
Spray Guns
All-PTFE Clean-In-Place sprayer for use with either pressurized or vacuum-assisted flask washing
  • All-PTFE construction
  • Sprayer consists of four, 3-inch sections to allow flexibility
  • Multiple extenders can be used with sprayer
PTFE Clean-In-Place sprayer with #15 Ace-Thred for use with bench-scale flasks in sizes 1000 - 6000 mL, and either pressurized or vacuum-assisted flask washing. ACE recommends that the sprayer ball be positioned around the mid-point of the vessel. Minimum flange size of 100 mm is required if installed through an angle joint.
Used with flasks ranging from 1000 - 6000 mL.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty