Would you like to log in again?
Tubing Connectors, Ace Glass Incorporated
Supplier: Ace Glass
5855-73 5855-10 5855-08 5855-71 5855-80
75877-586EA 85.98 USD InStock
75877-586 75877-578 75877-582 75877-584 75877-580
Tubing Connectors, Ace Glass Incorporated
Fittings and Connectors
Small tubing connector for use with Chromatography to tubing adapters
  • Separate PTFE nut, Kel-F wedge insert, and wedge insertion tool make up the complete connector
  • Connector components for either 1.5 mm or 2 mm I.D. tubing
  • Wedge insertion tool mechanically wedges the tubing between the insert and the inside wall of the nut
Small tubing connector comprised of a virgin TFE 1/4"–28 UNF 2B male nut and a Kel-F wedge insert. Use with ACE chromatograpy column to tubing filter adapters.
Suitable for vacuum or pressure applications, as tubing won't slip off or blow out.
Packaging: The PTFE nuts and Wedge Inserts are sold in packs of 12. The Wedge Insertion Tool is sold individually.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty