Would you like to log in again?
Two-Piece Pressure Reactor with Jacketed Flask, Ace Glass
Supplier: Ace Glass
6427-470 6427-464 6427-468 6427-466 6427-472 6427-474
75877-598EA 18139.11 USD InStock
75877-598 75877-588 75877-594 75877-596 75877-590 75877-592
Two-Piece Pressure Reactor with Jacketed Flask, Ace Glass
Laboratory Reactor Systems
Complete, two-piece pressure reactor system with jacketed flask, designed to perform low to moderate positive pressure reactions, synthesis and catalysis.
  • Jacketed flask made of borosilicate glass
  • Two-piece design allows the use of large agitators and facilitates product removal and clean-out
  • Reactor uses Ace-Thred inlet/outlets to achieve a leak-tight system
  • Rated 35psig at 100 degrees C
Complete pressure reactor system with two-piece design offers a flanged flask and head to allow for the use of large agitators, and to facilitate product removal and clean-out. The system includes a jacketed flask, a flanged multi-neck head, an addition funnel, a condenser, a pressure manifold, and a super magnet stirring assembly.
Ordering information: Stand and support shelf not included.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty