Would you like to log in again?
Scale-Up Series Bench-Top Reactor System, Ace Glass
Supplier: Ace Glass
6449-07 6449-04 6449-03 6449-10 6449-05 6449-06 6449-08 6449-09 6449-02
75877-624EA 7026.75 USD InStock
75877-624 75877-626 75877-620 75877-630 75877-622 75877-632 75877-628 75877-608 75877-618
Scale-Up Series Bench-Top Reactor System, Ace Glass
Laboratory Reactor Systems
Ace Glass Scale-Up Series plain bench-top reactor system includes the essential items needed to begin building a reactor for your specific application.
  • Flask made of high-quality borosilicate glass.
  • Reactors are sized for bench-top use, and their geometry translates into ACE Kilo Scale Reactors.
  • Base system includes a flask, a CAPFE o-ring, and a stand with supports.
Scale-Up Series reactors are available in 100 mL to 6000 mL capacities, and due to their geometry, the reaction can be translated into Kilo Scale reactors.
Ordering information: Base systems include the essential items, and can be customized by adding a head, condenser, funnel, heating mantle, or other items, which are sold separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty