Would you like to log in again?
Circulator Adapters, Ace Glass Incorporated
Supplier: Ace Glass
12187-70 12187-62 12187-81 12187-73 12187-61 12187-72 12187-74 12187-58 12187-82 12187-71 12187-63 12187-83 12187-84
75877-700EA 854.91 USD InStock
75877-700 75877-612 75877-702 75877-686 75877-696 75877-688 75877-698 75877-610 76107-048 75877-704 75877-692 75877-694 75877-690
Circulator Adapters, Ace Glass Incorporated
Fittings and Connectors
Stainless steel beaded pipe adapters designed for use with various manufacturers' circulators, and Ace Glass reaction systems.
  • Made of 304 stainless steel
  • Adapters with hose barbs connect to hoses on a circulator
  • Adapters with "M" style fittings connect to NPT threads
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty