Would you like to log in again?
Cell Viability Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
CQMT-500
75877-942EA 297.75 USD InStock
75877-942
Cell Viability Assay Kit, BioAssay Systems
Assays Cellular Assays
Homogeneous colorimetric assay for cell viability, proliferation, cytotoxcity and evaluation of anticancer agents.
  • Non-radioactive assay (cf. 3H-thymidine incorporation assay)
  • As low as 950 cells can be accurately quantified
  • High-throughput assay using 96-well plates allows processing tens of thousands of samples per day
  • No wash and reagent transfer steps are involved
The study of cell proliferation and cell viability requires the accurate quantification of the number of viable cells in a cell culture. Therefore, assays for calculating cell viability are necessary for optimizing cell culture conditions, evaluating cell growth factors and nutrients, discovering novel antibiotics and anti-cancer drugs, evaluating toxic effects of environmental pollutants and cell mediated toxicity and studying programmed cell death (apoptosis). The CellQuanti-MTT™ assay kit provides a convenient, sensitive, quantitative and reliable assay for determining the number of viable cells in a given culture. This homogeneous colorimetric assay is based on the conversion of a tetrazolium salt MTT, a pale yellow substrate, to formazan, a purple dye. This cellular reduction reaction involves the pyridine nucleotide cofactors NADH/NADPH and is only catalyzed by living cells. The formazan product has a low aqueous solubility and is present as purple crystals. Dissolving the resulting formazan with a solubilization buffer permits the convenient quantification of product formation. The intensity of the product color, measured at 550 - 620 nm, is directly proportional to the number of living cells in the culture. Reagents in the kit have been carefully formulated and optimized for sensitivity, assay robustness and automation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty