Would you like to log in again?
QuantiChrom™ Acetylcholinesterase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DACE-100
75877-948EA 565.83 USD InStock
75877-948
QuantiChrom™ Acetylcholinesterase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of acetylcholinesterase activity.
  • Detection range: 10 to 600 U/L AChE activity in 96-well plate assay
  • Can be readily automated as a high-throughput assay for thousands of samples per day
Acetylcholinesterase (EC 3.1.1.7, AChE), also known as RBC cholinesterase, is found primarily in the blood and neural synapses
BioAssay Systems’ QuantiChrom™ Acetylcholinesterase Assay is based on an improved Ellman method in which thiocholine produced by the action of acetylcholinesterase forms a yellow color with 5,5’-dithiobis(2-nitrobenzoic acid). The intensity of the product color, measured at 412 nm, is proportional to the enzyme activity in the sample.
AChE catalyzes the hydrolysis of the neurotransmitter acetylcholine into choline and acetic acid, a reaction necessary to allow a cholinergic neuron to return to its resting state after activation. Studies show serum cholinesterase activity may relate to insecticide exposure or to one of a number of variant genotypes. As a result, cholinesterase levels of cells and plasma are used as a guide in establishing safety precautions relative to exposure and contact, as well as a guide in determining the need for workers to be removed from areas of contact with the organic phosphate insecticides.Simple, direct and automation-ready procedures for measuring AChE activity are highly desirable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty