Would you like to log in again?
QuantiChrom™ Alcohol Dehydrogenase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DADH-100
75877-950EA 538.32 USD InStock
75877-950
QuantiChrom™ Alcohol Dehydrogenase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of alcohol dehydrogenase activity and evaluation of drug effects on its metabolism.
  • Linear detection range 0.4 to 80 U/L for 30 min reaction
  • Detection limit of 0.1 U/L for 120 min reaction
  • This homogeneous "mix-incubate-measure" type assay can be readily automated for processing thousands of samples per day
Alcohol dehydrogenase (ADH) is an oxidoreductase which catalyzes the interconversion of alcohols and aldehydes or ketones
BioAssay Systems’ non-radioactive, colorimetric ADH assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is directly proportional to the enzyme activity.
ADH is important in humans and other organisms for the break down of alcohols which may otherwise be toxic. In yeast and some bacteria, ADHs catalyze the opposite reaction and produce alcohol as part of fermentation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty