Would you like to log in again?
QuantiChrom™ Mannosidase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DAMA-100
75877-956EA 577.57 USD InStock
75877-956
QuantiChrom™ Mannosidase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of alpha-mannosidase enzyme activity.
  • Linear detection range 1 to 250 U/L for a 10-minute reaction
  • This homogeneous "mix-incubate-measure" type assay can be readily automated for processing thousands of samples per day
α-Mannosidase (AMA) is an enzyme which catalyzes the cleavage of the α form of mannose. α-Mannosidase assists in the breakdown of complex sugars from glycoproteins in the lysosome. Defective AMA or deficient AMA activity causes α-mannosidosis and leads to deterioration of the central nervous system in children. BioAssay Systems’ non-radioactive, colorimetric AMA assay is based on the cleavage of 4-nitrophenol from the synthetic substrate. Nitrophenol becomes intensely colored after addition of the stop reagent. The increase in absorbance at 405 nm after addition of the stop reagent is directly proportional to the enzyme activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty