Would you like to log in again?
QuantiChrom™ Glucose Dehydrogenase Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
DGDH-100
75877-972EA 525.51 USD InStock
75877-972
QuantiChrom™ Glucose Dehydrogenase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of D-glucose dehydrogenase enzyme activity and drug effects on its metabolism.
  • Linear detection range 0.5 to 200 U/L for 15 min reaction
  • Can be readily automated for processing thousands of samples per day
Glucose dehydrogenase (GDH) belongs to the family of oxioreductases, specifically those acting on the CH-OH group of donor with other acceptors. GDH participates in the pentose phosphate pathway. BioAssay Systems’ non-radioactive, colorimetric GDH assay is based on the reduction of the tetrazolium salt MTT in a NADH-coupled enzymatic reaction to a reduced form of MTT which exhibits an absorption maximum at 565 nm. The increase in absorbance at 565 nm is proportional to the enzyme activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty